Learn More
MP Biomedicals™ CoolTeenPrep™ Adapter for 6 x 15 mL Tube Holder on FastPrep-24
CoolTeenPrep™ Adapter for 6 x 15 mL Tube Holder on FastPrep-24 for cryogenic grinding and lysis on FastPrep-24.
Brand: MP Biomedicals™ 116002530
Code : 50
Additional Details : Weight : 0.01000kg
Description
CoolTeenPrep™ 6 X 15 mL Adapter is a cryogenic adapter which allows simultaneous lysis of 6 x 15 mL samples. Based on passive temperature control technology, the CoolTeenPrep™ adapter ensures an efficient cooling of the samples as dry-ice, placed into the moving tray, is in direct contact with the sample tubes. Due to a high heat transfer capacity and FastPrep™ precise settings of lysis parameters, the samples can be repeatably homogenized in their unthawed state. A wide range of disposable 15 mL Lysing Matrix tubes ensure thorough homogenization of any sample type in seconds.
- Application: CoolTeenPrep™ 6 X 15 mL Adapter is ideally suited for extractions of any temperature unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates, and enzymes from even the hardest samples to lyse.
- Capacity: 15 tubes
- Sample Volume: Up to 12 g
Specifications
Cryogenic Adapter | |
15°C to 30°C | |
6 x 15 mL | |
627 g |
12 g | |
Extractions of any temperature-unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates and Enzymes from even the hardest samples to lyse | |
1.382 lb. |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.