Learn More
MP Biomedicals™ HiPrep™ Adapter for 48 x 2 mL Tube Holder on FastPrep-24
HiPrep™ Adapter for FastPrep-24 instrument is a double decker adapter that holds 48 x 2 mL tubes for room temperature cell grinding and lysis.
Brand: MP Biomedicals™ 116002527
Code : 50
Additional Details : Weight : 0.01000kg
Description
Double decker adapter that holds 48 x 2 mL Lysing Matrix tubes. The ergonomic design of the adapter ensures easy loading of Lysing Matrix tubes that remain securely sealed during the homogenization. It stands stable on the bench top and is commonly used as a tube rack. Ideally suited for high-throughput applications, up 48 samples can be homogenized simultaneously. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapters and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
- Application Notes: The FastPrep™-24 Instrument is ideally suited for high-throughput applications.
- Capacity: 48 tubes
- Sample Volume: Up to 10 g
Specifications
High-throughput Adapter | |
15°C to 30°C | |
48 x 2 mL | |
467 g |
10 g | |
High-throughput applications, up to 24 and 48 samples can be homogenized simultaneously | |
1.029 lb. |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.