missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastDNA™ SPIN Kit for Soil, 50 mL Tubes

FastDNA SPIN Kit for Soil is used for extracting bacterial, fungal, plant, and animal DNA from soil and other environmental samples. It uses Lysing Matrix E for sample processing on a FastPrep™ instrument.

Brand:  MP Biomedicals™ 116560600

Code : NEW

Additional Details : Weight : 0.68000kg Transport : UN number : 1760 Chem class : 8 Pack group : III


Product Code. 15232830

  • £363.58 / Each
Estimated Shipment: 18-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Product Overview: The FastDNA Spin Kit for Soil is designed to isolate bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples. Soil is processed by a FastPrep™ instrument and Lysing Matrix E tubes designed to efficiently lyse all microorganisms. The released DNA is purified by a silica-based spin filter method and is suitable for PCR analysis and other downstream applications. Complete lysis of all organisms, including historically difficult sources such as eubacterial spores and endospores, gram positive bacteria, yeast, algae, nematodes and fungi is achieved within seconds using a FastPrep instrument and Lysing Matrix E tubes.
  • Applications: Cited in over 6000 scientific publications, the FastDNA Spin Kit for Soil delivers the highest DNA yield from any environmental sample including soil, sediments, sludge, compost, manure, rhizosphere or waste water. The FastDNA Spin Kit for Soil contains Lysing Matrix tubes and all the reagents needed to extract DNA in a process that efficiently removes humic acids and other PCR inhibitors
  • Sample Type: Seeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants, Roots, Soil, Wastewater, Yeast
  • Sample Volume: Approximately 200 μL to 50 mL; up to approximately 25 g
Specifications

Specifications

Room Temperature
Use to isolate bacterial, fungi, plant and animal genomic DNA from soil and environmental samples
FastDNA-24 Kits
Seeds, Spores, Animal Tissues, Feces, Food, Fungi, Plants, Roots
10 each Garnet Lysing Matrix Tubes (50 mL), 60 mL PLS Solution, 15 mL MT Buffer, 350 mL Sodium Phosphate Buffer, 25 mL PPS for Soil, 120 mL Binding Matrix, 120 mL GTC Wash Buffer, 12 mL SEWS-M, 60 mL TES, 200μL BBS Gel Loading Dye, 10 each SPIN Filter Tubes (50 mL)
Approximately 200 μL to 50 mL; Up to approximately 25 g
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : guanidine thiocyanate, sodium dodecyl sulphate, polyvinylpyrrolidone

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.