Learn More
MP Biomedicals™ FastDNA™-96 Plant/Seed DNA Kit
FastDNA™-96 Plant/Seed DNA Kit is used for rapid, high-throughput isolation of PCR-ready genomic DNA from plants tissues and seeds. For use with the FastPrep-96™ Instrument.
Brand: MP Biomedicals™ 119696600
Code : NEW
Additional Details : Weight : 0.68000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- Product Overview: Rapid, High-Throughput Isolation of PCR-Ready Genomic DNA from Plant and Seed Samples The FastDNA™-96 Plant and Seed DNA Kit is designed for the simple and rapid isolation of PCR-quality DNA from vegetative samples in approximately 50 minutes. Plant samples including stems, roots, leaves, buds, flowers, fruits and seeds are efficiently lysed in approximately 60 seconds with the FastPrep-96™ Instrument. Up to 80 mg samples are placed into each 1.2 mL tube of a FastDNA-96™ Lysing Matrix Rack (96-deep well plate) containing 2.0 mm specially stabilized Zirconium oxide lysing particles, which are designed to efficiently lyse a wide variety of plant specimens, while in the presence of a specially formulated lysis solution. The cleared lysate is then applied to a binding matrix to isolate the DNA, and spin column plates are used to simplify liquid handling. Eluted DNA is then applied to the MP-96 Inhibitor Removal Plate which removes tannins, polyphenols and polysaccharides that can inhibit PCR. Purified DNA is eluted in an EDTA-free, DNA elution solution, and is ready for downstream applications including digestion, electrophoresis, arrays and PCR (A260/A280 ratios ≥1.8). Yield is typically 5 μg of total DNA eluted in 50 to 100 μL of elution solution. This procedure can be performed in minutes, and there is no need for organic denaturants or proteinases.
- Applications: The FastDNA™-96 Plant/Seed DNA Kit for FastPrep™-96 is used for the simple, rapid isolation of inhibitor-free, PCR-quality DNA from a variety of plant sample sources including leaves, stems, buds, flowers, fruit, and seeds. Eluted DNA can be used for downstream molecular-based applications including PCR, arrays, digestion, electrophoresis etc.
- Sample Source: Plant Tissues
- Sample Type: Seeds, Plants, Roots, Buds, Flowers, Fruits, Leaves, Stems
- Sample Volume: Up to 80 mg
Specifications
Room Temperature | |
Seeds, Plants, Roots, Buds, Flowers, Fruits, Leaves, Stems | |
FastDNA-96 Kits | |
Up to 80 mg |
Kit | |
2 x 96 Preps | |
1 Ea. | |
Plant Tissues |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.