Learn More
MP Biomedicals™ FastDNA™-96 Soil Microbe DNA Extraction Kit
FastDNA™-96 Soil Microbe DNA Extraction Kit is designed to quickly and efficiently isolate humic-free, PCR-ready genomic DNA from a variety of soil samples.
Brand: MP Biomedicals™ 119696200
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Product Overview: The FastDNA-96™ Soil Microbe DNA Kit quickly and efficiently isolates humic-free, PCR-ready genomic DNA from soil samples in approximately 50 minutes. Gram(+/-) bacteria, fungi, plant and animal tissues, algae, spores and many other members of a soil population including clay, sandy, silty, peaty, chalky, and loamy soils, are efficiently lysed in approximately 60 seconds with the FastPrep-96™ Instrument.
- Sample Type: Feces, Soil, Wastewater, Environmental Water, Sludge
Specifications
Kit | |
2 x 96 Preps | |
1 Ea. |
Feces, Soil, Wastewater, Environmental Water, Sludge | |
FastDNA-96 Kits |
Safety and Handling
- FastDNA-96 Soil Microbe DNA Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : isopropanol, ethanol, glycerol, guanidine hydrochlride
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.