Learn More
MP Biomedicals™ FastDNA™ Kit
FastDNA™ Kit quickly and efficiently isolates high quality genomic DNA from plants, animals, bacteria, yeast, algae, and fungi.
Brand: MP Biomedicals™ 116540400
464.23 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 1.25584kg Transport : UN number : 2922 Chem class : 8 Pack group : III
Description
- Uses Lysing Matrix A (garnet and 1 1/4 inch ceramic bead) for cell lysis and a silica-based method for the purification process.
- Lysing Matrix A and 3 different chaotropic buffers are provided for the homogenization of a wide variety of sample types (plant and animal tissues, cultured cells, bacteria, yeast, fungi, insects, etc.).
- DNA is released within 40 seconds, and is purified by a silica-based procedure.
- Eluted DNA is ready for digestion, electrophoresis, PCR and any other desired application.
Specifications
Kit | |
Used to isolate DNA from virtually any source | |
100 Preps |
Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Yeast, Algae, Cultured Cells, Insects | |
Lysing Matrix A 100 2 mL tubes, 1/4 Ceramic Spheres 100 spheres, Binding Matrix 66 mL, Concentrated SEWS-M 12 mL, DES 25 mL, CLS VF 90 mL, PPS 25 mL, CLS-TC 110 mL, CLS-Y 110 mL | |
FastDNA Kit |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H301-Toxic if swallowed.
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash hands thoroughly after handling.
- P273-Avoid release to the environment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ first aider if you feel unwell.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor / first aider
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ first aider if you feel unwell.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor / first aider
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P330-Rinse mouth.
- P321-Specific treatment (see on this label).
- P310-Immediately call a POISON CENTER/doctor/first aider.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : acetic acid
Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.