Learn More
MP Biomedicals™ FastDNA™ SPIN Kit
FastDNA™ SPIN Kit is used for isolation of DNA from up to 200 mg of almost any sample in less than 30 minutes. This is the updated version of the well-known FASTDNA Kit and can be used with any FastPrep Instrument.
Brand: MP Biomedicals™ 116540600
717.64 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : LE
Additional Details : Weight : 1.46880kg Transport : UN number : 1760 Chem class : 8 Pack group : III
Requires : FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge that can freely spin 2.0 mL tubes, Microcentrifuge tubes (2.0 mL and 1.5 mL), Rotator or low-speed vortex
Description
- Product Overview: The FastDNA™ SPIN Kit, an updated version of the well-known FastDNA™ Kit, is used with any FastPrep™ Instrument to lyse and subsequently isolate DNA from up to 200 mg of almost any sample in less than 30 minutes.
- Features: Isolation of genomic DNA from plants, animals, bacteria, yeast, algae, and fungi using a silica spin fi lter method: Rapid and reproducible sample lysis with the FastPrep™-24 or FastPrep™ FP120 Instrument, Lyse and isolate DNA in less than 30 minutes from a variety of sample types, Flexible format eliminates need for separate DNA isolation kits, No hazardous organic reagents required, SPIN Filters are included to streamLine silica handling.
- Applications: The Fast DNA™ SPIN Kit, an updated version of the well known Fast DNA TM Kit, is used with the FastPrep™-24 or FastPrep™ FP120 Instrument to lyse and subsequently isolate DNA from up to 200 mg of almost any sample in less than 30 minutes.
- Sample Type: Seeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants, Roots, Wastewater, Yeast
- Sample Volume: Up to 200 mg per prep
Specifications
Seeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants | |
Lysing Matrix A 100 x 2 mL Tubes, 100 x 0.25 in. Ceramic Spheres, 66 mL Binding Matrix, 12 mL Concentrated SEWS-M, 25 mL DES, 90 mL CLS-VF, 25 mL PPS, 110 mL CLS-TC, 110 mL CLS-Y, 100 x SPIN modules, 100 x Catch Tubes, 1 x User Manual, 1 x MSDS | |
FastDNA-24 Kits |
Isolates genomic DNA from plants, animals, bacteria, yeast, algae and fungi | |
100 Preps | |
Up to 200 mg per prep |
Safety and Handling
- Fast DNA Spin Kit
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H301-Toxic if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- P264-Wash thoroughly after handling.
- P321-Specific treatment (see on this label).
- P270-Do not eat, drink or smoke when using this product.
- P330-Rinse mouth.
Supplemental information
- MIXTURE LIST-Contains : Sodium Dodecyl Sulfate, Sodium Dodecyl Sulfate, Sodium Dodecyl Sulfate, (R*, R*)-(±)-1, 4-dimercapto
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.