Learn More
MP Biomedicals™ FastDNA™ SPIN Kit for Plant and Animals Tissues
FastDNA™ SPIN Kit for Plant and Animals Tissues is used to isolate DNA from plant and animal tissues.
Brand: MP Biomedicals™ 116540800
Code : NEW
Additional Details : Weight : 0.25000kg Transport : UN number : 1760 Chem class : 8 Pack group : III
Requires : FastPrep™ Instrument or alternative high speed disruptor; Microcentrifuge that can freely spin 2.0 mL tubes; Microcentrifuge tubes (2.0 mL and 1.5 mL); Rotator or low-speed vortex
Description
- Product Overview: The FastDNA™ Spin Kit for Plant and Animals Tissues quickly and efficiently isolates high quality genomic DNA from plant and animal tissues using Lysing Matrix D (1.4 mm ceramic beads) for cell lysis and a silica-based spin filter method for the purification process.
- Sample Type: Animal Tissues, Plants, Cultured Cells
Specifications
Animal Tissues, Plants, Cultured Cells | |
100 x 2 mL Tubes Lysing matrix D, 0.25 in. Ceramic Spheres, 100 mL Binding Matrix, 12 mL Concentrated SEWS-M, 25 mL DES, 90 mL CLS-VF, 25 mL PPS, 110 mL CLS-TC, 100 x SPIN Modules, 100 x Catch Tubes | |
FastDNA-24 Kits |
Use to isolate DNA from plant and Animal tissues | |
100 Preps |
Safety and Handling
- FastDNA Green SPIN Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium dodecyl sulphate, guanidium thioyanate
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.