missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastPrep-96™ High-throughput Bead Beating Grinder and Lysis System

FastPrep-96™ high-throughput bead beating grinder and lysis system is a high-speed, digitally controlled benchtop homogenizer for the lysis of biological samples. It can process up to 192 samples in 2 x 96 deep well plates. It can perform 1800 oscillations/min and 1.5-inch stroke.

Brand:  MP Biomedicals™ 116010500

Product Code. 15422253

  • £19075.00 / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Product Overview: MP Biomedicals' FastPrep-96™ is a versatile high-speed homogenizer offering the ultimate in high-throughput sample preparation. Developed for resistant samples, the FastPrep-96™ instrument uses a high-speed linear motion to disrupt thoroughly any tissues and cells in just seconds through the simultaneous beating of specialized Lysing Matrix beads on the sample material. This high-performance system saves hours of work during the sample preparation stage and allows for the purification of high yields of intact DNA, RNA, proteins and other metabolites.

  • Service: One-year warranty is included with purchase. For extended warranty, please view the MPCare FastPrep-96™ Service Agreement.
  • Application: FastPrep™-96 is used to homogenize biological samples. FastPrep™-96 Instrument lyses thoroughly and quickly any tissues and cells and thus allows easy and reproducible isolation of stable RNA, active proteins and full-length genomic DNA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

66 x 44 x 70 cm (26 x 17.3 x 27.5 in.)
Built-in adapter
1 to 60 sec.
2°C to 48°C
49 kg
110/220V
Ideal for high throughput sample homogenization, grinding, lysis and preparation, nucleic acid isolation and purification
<65 dBA
35°F to 100°F
108 lb.
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.