Learn More
MP Biomedicals™ FastRNA™ Pro Blue Kit, 2 mL Tubes
FastRNA™ Pro Blue Kit, 2 mL Tubes are used for total RNA isolation from up to 1010 gram positive and gram negative bacteria. Uses Lysing Matrix B for cell lysis. For use with the FastPrep-24™ or FastPrep™ FP120 Instrument.
Brand: MP Biomedicals™ 116025050
406.14 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 80
Additional Details : Weight : 0.46170kg Transport : UN number : 2810 Chem class : 6.1 Pack group : II
Requires :
- FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, RNase Erase™
- Recommended: Chloroform, 100% ethanol, 75% ethanol, 1.5 or 2.0 mL RNase-free microcentrifuge tubes, Agarose, Gel loading dye and RNA size marker
Description
- Product Overview: The FastRNA™ Pro Blue Kit is designed to efficiently isolate total RNA from gram positive and gram negative bacteria. Using the FastPrep™-24 or FastPrep™ FP120 Instrument, up to 1010 cells are homogenized by Lysing Matrix B in impact-resistant 2 mL tubes. Total RNA is released into the proprietary, protective RNApro™ Solution.
- Applications: FastRNA™ Pro Blue Kit is used to isolate total RNA from gram positive and gram negative bacteria. Isolated RNA is suitable for RT-PCR analysis and other downstream applications.
- Sample Source: Microorganisms
- Sample Type: Bacteria (Gram +/-)
- Sample Volume: Up to 500 mg per prep
Specifications
Bacteria (Gram +/-) | |
RNApro™ Solution (1 x 55 mL Bottle), DEPC-H2O (1 x 15 mL Bottle), Lysing Matrix B (50 x 2 mL Tubes), Short Protocol, User Manual, MSDS, Certificate of Analysis | |
FastRNA-24 Kits | |
Microorganisms |
RT-PCR analysis and other downstream applications | |
50 Preps | |
50 x 2 mL |
Safety and Handling
- FastRNA Pro Blue Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Phenol, Guanidine Thiocyanate
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.