Learn More
MP Biomedicals™ FastRNA™ Pro Soil-Direct Kit
FastRNA™ Pro Soil-Direct Kit is designed to efficiently isolate total RNA from organic material found in soil samples. Suitable for RT-PCR.
Brand: MP Biomedicals™ 116070050
714.11 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : Z3
Additional Details : CAS Number : 593-84-0 Weight : 0.50000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Requires : RNase Erase™ recommended, FastPrep™ Instrument or alternative high speed disruptor, 100% Ethanol, Microcentrifuge, Chilled 70% Ethanol (prepared with DEPC-treated H2O), Chilled Isopropanol, LRNase-free Microcentrifuge
Description
- Product Overview: The FastRNA™ Pro Soil-Direct Kit is designed to efficiently isolate total RNA from organic material found in soil samples.
- Application: FastRNA™ Pro Soil-Direct Kit is used to isolate total RNA from organic material found in soil samples. Total RNA isolated is suitable for RT-PCR analysis and other downstream applications.
- Sample Source: Microorganisms
- Sample Type: Feces, Soil, Wastewater, Environmental Water, Sludge
- Sample Volume: Up to 500 mg per prep
Specifications
Feces, Soil, Wastewater, Environmental Water, Sludge | |
Soil Lysis Solution, (1:1) Phenol: Chloroform, Inhibitor Removal Solution, RNAMATRIX™, Slurry, Binding Solution, Wash Solution Concentrate, DEPC-treated Water, Lysing Matrix E 50 | |
Lysing Matrix E | |
50 x 2 mL |
Nucleic Acid isolation and purification, Protein isolation, molecular biology, gene expression, microbiological analysis | |
50 Preps | |
FastRNA-24 Kits |
Safety and Handling
- fast RNA pro soil-direct kit
Signal Word
- Danger
Hazard Category
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 1 B
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 4
- Germ cell mutagenicity Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H301-Toxic if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H312-Harmful in contact with skin.
- H311-Toxic in contact with skin.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : guanidine thiocyanate, phenol, chloroform, phenol, isoamyl alohol, acetic acid,diethyl pyrocarbonat
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.