Learn More
MP Biomedicals™ FastRNA™ Pro Soil-Indirect Kit
FastRNA™ Pro Soil-Indirect Kit is used to isolate total RNA from organic material found in soil samples. Total RNA isolated is suitable for RT-PCR analysis and other downstream applications.
Brand: MP Biomedicals™ 116075050
536.73 GBP valid until 2024-12-31
Use promo code "21969" to get your promotional price.
Description
- This kit allows for the sample to be slurried with water or buffer in order to further separate biomass from the soil or environmental matrix, and the resulting supernatant is then filtered, lysed, and purified through the rest of the protocol.
- Total RNA isolated is suitable for RT-PCR analysis and other downstream applications.
Specifications
Feces, Soil, Wastewater, Sludge | |
Kit | |
Up to 500 mg per prep |
50 Preps | |
2 mL | |
Microorganisms |
Safety and Handling
- FastRNA PRO SOIL - INDIRECT KIT
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H332-Harmful if inhaled.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : guanidium thiocyanate
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.