Learn More
MP Biomedicals™ FastRNA™ SPIN Kit for Yeast
FastRNA™ SPIN Kit for Yeast quickly and efficiently isolates high-quality, total RNA from tough-to-lyse yeast strains, fungi and algae using a specialized Lysing Matrix (Yttria-stabilized zirconium oxide, 0.5 mm) for cell lysis and SPIN columns for the purification process.
Brand: MP Biomedicals™ 116030050
413.88 GBP valid until 2024-12-31
Use promo code "21969" to get your promotional price.
Alert:
Personal Protective Equipment: Respiratory protection, hand protection and eye protection. Take the usual safety precautions while handling the product will provide adequate protection against this potential effect.
Requires : FastPrep Instrument or alternative high speed disruptor, Microcentrifuge that can spin up to 14,000 rpm, 100% Ethanol (or 95% Ethanol)
This item is not returnable. View return policy
Description
- Product Overview: The FastRNA™ SPIN Kit for Yeast quickly and efficiently isolates high-quality, total RNA from tough-to-lyse yeast strains, fungi, and algae in approximately 15 minutes using a specialized Lysing Matrix (Yttria-stabilized zirconium oxide, 0.5 mm) for cell lysis and SPIN columns for the purification process.
- Sample Type: Spores, fungi, yeast, algae
Specifications
Nucleic Acid Isolation and Purification | |
50 Preps | |
FastRNA-24 Kits |
Lysing matrix Y (0.5 mm yttria-stabilized ZrO2) 50 x 2 mL tubes, FastRNA Lysis Buffer 50 mL, FastRNA prep Buffer 25 mL, FastRNA Wash Buffer 12 mL, Water (DNase/RNase-free) 1 mL, FastRNA spin column 1 50 each, FastRNA spin column 2 50 each, collection tubes 2 mL | |
Lysing Matrix Y | |
50 x 2 mL |
Safety and Handling
- FastRNA Lysis Buffer
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Guanidine Thiocyanate, TRITON X-100, N-Lauroylsarcosine Sodium Salt
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.