Learn More
MP Biomedicals™ FastRNA™ SPIN Kit for Microbes
FastRNA™ SPIN Kit for Microbes isolates total RNA from bacterial cell cultures and other microbial sources in approximately 15 minutes. Does not use phenol or reducing agents.
Brand: MP Biomedicals™ 116020050
529.98 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.59400kg Transport : UN number : 2922 Chem class : 8 Pack group : II
Alert:
Research Use Only
Description
- Product Overview: The FastRNA™ SPIN Kit for Microbes quickly and efficiently isolates high-quality, total RNA from tough-to-lyse bacterial cell cultures and other microbial sources in approximately 15 minutes using a zirconium silicate Lysing Matrix (0.1 mm) to lyse bacteria and SPIN columns for the purification process.
- Sample Source: Microorganisms
- Sample Type: Bacteria (Gram +/-)
Specifications
Room Temperature | |
Lysing Matrix B (0.1 mm Zirconium Silicate), 50 x 2 mL Tubes, 50 mL FastRNA Lysis Buffer, 25 mL FastRNA Prep Buffer, 12 mL FastRNA Wash Buffer, 1 mL Water (DNase/RNase-free), 50 Tubes FastRNA Spin Column 1, 50 Tubes FastRNA Spin Column 2, 100 x 2 mL Collection Tubes | |
FastRNA-24 Kits |
Nucleic Acid Isolation and Purification | |
50 Preps |
Safety and Handling
- FastRNA SPIN Kit for Microbes
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P320-Specific treatment is urgent (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : guanidine hydrochloride, triton X-100
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.