missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ficin

From fig tree

Brand:  MP Biomedicals™ 0219876180

 View more versions of this product

Product Code. 11446831

  • £1019.72 / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Activity: 300-500 milk clotting units/mg. Hygroscopic.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
Cleavage of Mouse IgG
Fig tree Ficus glabrata
Ficin
Iodoacetamide, Iodoacetic Acid, N-ethylmaleimide, Mercuric Chloride, DFP (Diisopropyl Fluorophosphate), TLCK (Na-p-Tosyl-lysine Chloromethyl Ketone), TPCK (N-Tosyl-L-phenylalanine Chloromethyl Ketone)
5 to 7.5
4°C
100 g
≈25
Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers