missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Glucose Oxidase
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 kU
250 kU
50 kU
Unit Size:
10ku
250ku
50ku
Description
Type X. Activity: >100U/mg. Unit Definition: one unit oxidizes 1.0μmol of β-D-glucose to D-gluconic acid and H2O2 per minute at 35°C, pH 5.1.
Specifications
Specifications
| Concentration | >100 U/mg |
| Color | Yellow |
| pH | 4 to 7 |
| For Use With (Application) | Glucose Oxidase can be utilized in the enzymatic determination of D-glucose in solution |
| Content And Storage | -20°C |
| Source | Aspergillus niger |
| Quantity | 250 kU |
| Product Type | Glucose Oxidase |
| Molecular Weight (g/mol) | ≈160000 |
| Inhibitors | Ag+, Hg2+, Cu2+ , 4-chloromercuribenzoate, 50% D-arabinose |
| Show More |
Safety and Handling
Product Identifier
- Glucose Oxidase
- Danger
- Respiratory sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Glucose Oxidase
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction