missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Glucose Oxidase

From Aspergillus niger

Brand:  MP Biomedicals™ 0219519610

75.18 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.



 View more versions of this product

Product Code. 11481092

  • £89.50 / 10ku

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Type X. Activity: >100U/mg. Unit Definition: one unit oxidizes 1.0μmol of β-D-glucose to D-gluconic acid and H2O2 per minute at 35°C, pH 5.1.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

>100 U/mg
4 to 7
-20°C
10 kU
≈160000
Lyophilized Powder
Yellow
Glucose Oxidase can be utilized in the enzymatic determination of D-glucose in solution
Aspergillus niger
Glucose Oxidase
Ag+, Hg2+, Cu2+ , 4-chloromercuribenzoate, 50% D-arabinose
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : glucose oxidase

SDS
Documents

Documents

Certificates
Special Offers

Special Offers