missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ham's F-10 Medium
Optimized for clonal isolation and growth of CHO cells
Brand: MP Biomedicals™ 091440049
30.31 GBP valid until 2024-12-31
Use promo code "21941" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
4°C | |
Sterile | |
5.5 to 6.1 |
Ham's F-10 Medium | |
No Glutamine, No Sodium Bicarbonate | |
100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction