missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ 1X Hanks' Balanced Salt Solution, Modified
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
Used for washing cells and tissues while maintaining cells in a viable state. Buffered with phosphate and maintains a physiological pH. Contains phenol red.
Specifications
Specifications
| Chemical Name or Material | Hanks' Balanced Salt Solution (HBSS) |
| Color | Red |
| Concentration | 1 X |
| pH | 6.9 to 7.2 |
| Physical Form | Liquid |
| Quantity | 500 mL |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction