Learn More
MP Biomedicals™ Immunidiffusion (RID) Plates
Used for double and single radial immunodiffusion techniques
Brand:  MP Biomedicals™ 08421651
Product Code. 11403626
  • £104.00 / Pack of 12
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polystyrene construction with close-fitting covers to minimize gel dehydration
  • Comes with recessed bottoms to prevent scratching on bench surfaces
  • Hydrophilic in nature for easy gel pouring and better adherence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polystyrene
Double and single radial immunodiffusion techniques
15° to 30°C
Immunodiffusion Plate
12 Pack