missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lactase from Aspergillus oryzae
Cleaves lactose into its monosaccharide components, glucose and galactose
£160.06
Specifications
Concentration | >5000 U/g |
---|---|
Color | Off-White |
pH | 4 to 8 |
For Use With (Application) | Screen for blue white colonies in MCS of various plasmid vectors in E. coli or other Bacteria and to break down Lactose in milk |
Content And Storage | 4°C |
Description
- Hydrolyzes Lactose (4-o-β-D-Galactopyranosyl-D-glucose) and produces β-D-Galactose and α-D-Glucose
- Catalyses transglycosylation of glucose into allolactose, the inducer of β-galactosidase
- EC Number: 232-864-1
- MDL Number: MFCD00130623
- Format: Off white powder
- Specific Activity: >5000U/g
- Stability: Stable in pH range of 4.0 to 8.0
- Stable to heat
- Soluble in water, insoluble in ethanol, acetone and isopropyl alcohol
- Optimum pH is 4.5 and 4.8 when O-Nitrophenyl-beta-D- galactopyranoside and Lactose are used as substrate
Used to screen for blue white colonies in MCS of various plasmid vectors in Escherichia coli or other bacteria and to break down lactose in milk to make it suitable for people with lactose intolerance.
Specifications
>5000 U/g | |
4 to 8 | |
4°C | |
Lactase |
Off-White | |
Screen for blue white colonies in MCS of various plasmid vectors in E. coli or other Bacteria and to break down Lactose in milk | |
Aspergillus oryzae | |
Powder |