missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ LymphoSep™ Lymphocyte Separation Medium

LymphoSep™ Lymphocyte Separation Medium is a sterile, iso-osmotic, low viscosity solution for density centrifugation yielding a clear separation of lymphocytes from heparinized or defibrinated human blood.

Brand:  MP Biomedicals™ 091692249


 View more versions of this product

Product Code. 11444815

  • £72.05 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Lymphocytes are isolated by layering diluted, heparinized blood over a solution of sodium metrizoate that contains a polysaccharide
  • Polysaccharide aggregates the erythrocytes so that they can be sedimented by low centrifugation
  • The less dense lymphocytes remain as a clearly defined band at the plasma Lymphosep interface
  • Provides:
    • In vitro isolation of lymphocytes from diluted whole blood
    • Maximum yield of mononuclear cells
    • Easy one-step centrifugation
    • >96% cell viability of lymphocytes
    • Low endotoxins
    • Autoclave sterilization
TRUSTED_SUSTAINABILITY
Specifications

Specifications

15° to 30°C
Sterile
100 mL
Lymphosep Lymphocyte Separation Medium
6 to 9
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Lymphosep - Lymphocyte Separation Medium

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium diatrizoate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers