missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ TeenPrep™ Lysing Matrix D Tubes

TeenPrep™ Lysing Matrix D Tubes contain 1.4 mm zirconium-silicate spheres and are used to grind soft animal and plant tissues, cultured cells and insects/worms.

Brand:  MP Biomedicals™ 116983001

 View more versions of this product

Product Code. 12323873

  • £188.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Lysing Matrix D, 96-tube rack comes with 1.2 mL cluster-strip tube and cap assemblies. Each tube is pre-filled with 1.4 mm zirconium-silicate spheres and will not bind nucleic acids. Use with FastPrep-96 instrument or similar bead beater.

  • Tube size: 96-tube rack
  • Sample Types: Soft tissues such as brain, liver, kidney, lung and spleen; plant tissues such as leaves, roots and fruits; cultured cells; whole insects; nematodes
  • Matrix Composition: 1.4 mm zirconium-silicate spheres
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.4 mm Zirconium-silicate Spheres
Dnase-free
Nucleic Acid Isolation and Purification, Homogenization
Lysing Matrix D
Rnase-free
Animal Tissues, Plants, Cultured Cells, Insects, Worms
FastPrep-96 or similar bead beater
1.2 mL Cluster-strip Tube and Cap Assemblies
1 x 96-Well Plate
Less than 50 mg per prep
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ TeenPrep™ Lysing Matrix D Tubes > Lysing Matrix D, 96-tube Rack; Quantity: 1 x 96-Well Plate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.