Learn More
MP Biomedicals™ TeenPrep™ Lysing Matrix F, 15 mL Tubes
TeenPrep™ Lysing Matrix F, 15 mL Tubes are used to isolate RNA and protein from samples. Lysing Matrix F is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.
Brand: MP Biomedicals™ 116935025
Code : NEW
Additional Details : Weight : 0.32000kg
Description
- Impact-resistant 15 mL tubes contain 1.6 mm aluminum oxide particles and 1.6 mm silicon carbide particles
- With white caps
- Specific Gravity: Aluminum Oxide, 3.95; Silicon Carbide, 3.20
- Nucleic Acid Isolation and Purification
Specifications
15 mL | |
Dnase-free | |
Lysing matrix F | |
25 x 15 mL Tubes |
Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Coral Emulsions, Mold | |
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | |
Aluminum Oxide, Al2O3; Silicon Carbide, SiC | |
RNase-free |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.