missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix F, 2 mL Tubes
Lysing Matrix F, 2 mL Tubes are used to isolate RNA and protein from samples. Lysing Matrix F is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.
£253.34 - £342.38
Specifications
Sample Type | Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Coral Emulsions, Mold |
---|---|
DNase | Dnase-free |
For Use With (Application) | Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis |
Matrix | Lysing matrix F |
Molecular Formula | Aluminum Oxide, Al2O3; Silicon Carbide, SiC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11482420
|
MP Biomedicals™
116915050 |
50 x 2 mL Tubes |
£253.34
Pack of 50 |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11492420
|
MP Biomedicals™
116915100 |
100 x 2 mL Tubes |
£342.38
Pack of 100 |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Impact-resistant 2 mL tubes contain 1.6 mm aluminum oxide particles and 1.6 mm silicon carbide particles
- With white caps
- Specific Gravity: Aluminum Oxide, 3.95; Silicon Carbide, 3.20
- Nucleic Acid Isolation and Purification
Specifications
Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Coral Emulsions, Mold | |
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | |
Aluminum Oxide, Al2O3; Silicon Carbide, SiC |
Dnase-free | |
Lysing matrix F | |
RNase-free |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.