missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix H
Lysing Matrix H used to isolate RNA and Proteins from bacterial aggregates, biofilms, and whole insects.
£253.34 - £1370.58
Specifications
Capacity | 2 mL |
---|---|
Sample Type | Seeds, Animal Tissues, Plants, Soil, Bacteria, Ancient and Dried Samples, Clay, Wood |
DNase | Dnase-free |
Matrix | Lysing matrix H |
RNase | RNase-free |
Product Code | Brand | For Use With (Application) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Quantity | Price | Quantity & Availability | ||||
11422430
|
MP Biomedicals™
116917050 |
Nucleic Acid isolation and purification | 50 x 2 mL Tubes |
£253.34
Pack of 50 |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11432430
|
MP Biomedicals™
116917100 |
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | 100 x 2 mL Tubes |
£400.68
Pack of 100 |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15444683
|
MP Biomedicals™
116917500 |
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | 500 x 2 mL Tubes |
£1370.58
Pack of 500 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Impact-resistant 2 mL tube contains 2 mm glass beads and 2 mm yellow zirconium oxide beads
- With yellow caps
- Specific Gravity: Glass Beads, 2 mm; Yellow Zirconium Oxide Beads, 5.5
- Applications: Nucleic Acid Isolation and Purification
Specifications
2 mL | |
Dnase-free | |
RNase-free |
Seeds, Animal Tissues, Plants, Soil, Bacteria, Ancient and Dried Samples, Clay, Wood | |
Lysing matrix H |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.