missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix I
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 x 2 mL Tubes
50 x 2 mL Tubes
Unit Size:
Pack of 100
Pack of 50
Description
- Tube Size: 2 mL
- Sample Type: Plant and animal tissues, wood, seeds, very dense soils and clays, whole insects, ticks, ancient and dried samples, dry grinding fungal spores/rusts
- Molecule to be Enriched: DNA, RNA, proteins, other small molecules
- Matrix Composition: Matrix I, contains 2 mm yellow zirconia beads and a 4 mm black ceramic bead
Specifications
Specifications
| Sample Type | Seeds, Spores, Animal Tissues, Plants, Soil, Ancient and Dried Samples, Clay, Wood |
| DNase | Dnase-free |
| For Use With (Application) | Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis |
| Matrix | Lysing matrix I |
| Quantity | 50 x 2 mL Tubes |
| RNase | RNase-free |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Lysing Matrix I >
Spot an opportunity for improvement?Share a Content Correction