missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Poly-D-Lysine Hydrobromide Attachment Factors
Facilitate attachment, adherence, and spreading in the culture of adherent cells
£93.35 - £202.00
Specifications
Content And Storage | -20°C |
---|---|
Product Type | Poly-D-Lysine |
Sterility | Non-sterile |
Source | Synthetic |
Product Code | Brand | Product Type | Quantity | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Sterility | Price | Quantity & Availability | |||
11446632
|
MP Biomedicals™
0215017525 |
Poly-D-Lysine | 25 mg | Non-sterile |
£103.00
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11436632
|
MP Biomedicals™
0215017510 |
Poly-D-Lysine | 10 mg | Non-sterile |
£93.35
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11456632
|
MP Biomedicals™
0215017550 |
Poly-D-Lysine | 50 mg | Non-sterile |
£202.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Nonsterile.
Specifications
-20°C | |
Poly-D-Lysine | |
Non-sterile | |
Synthetic |