missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Poly-L-Lysine Hydrobromide Attachment Factors: 30,000-70,000 MW
Facilitate attachment, adherence, and spreading in the culture of adherent cells
£67.73
Specifications
Content And Storage | -20°C |
---|---|
Product Type | Poly-L-Lysine |
Source | Synthetic |
Product Code | Brand | Product Type | Quantity | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Sterility | Price | Quantity & Availability | |||
11440812
|
MP Biomedicals™
0219454305 |
Poly-L-Lysine | 5 mg | Sterile |
£67.73
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
Available nonsterile or gamma-irradiated.
Specifications
-20°C | |
Poly-L-Lysine | |
Synthetic |