missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Poly-L-Lysine Hydrobromide Attachment Factors: ≥70,000 MW
Facilitate attachment, adherence, and spreading in the culture of adherent cells
£404.00
Specifications
Content And Storage | -20°C |
---|---|
Product Type | Poly-L-Lysine |
Source | Synthetic |
Product Code | Brand | Product Type | Sterility | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Sterility | Quantity | Price | Quantity & Availability | |||
11446642
|
MP Biomedicals™
0215017780 |
Poly-L-Lysine | Non-sterile | 100 mg |
£404.00
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Available nonsterile or gamma-irradiated.
Specifications
-20°C | |
Poly-L-Lysine | |
Synthetic |