missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Protease, from S. aureus strain V8

Used for selective cleavage of proteins for amino acid sequence determination or peptide mapping

Brand:  MP Biomedicals™ 0215197201


 View more versions of this product

Product Code. 11414211

  • £217.00 / 1mg
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Unit Definition: One unit will hydrolyze casein to change A280 0.001 per minute, pH 7.8, at 37°C.
  • Extinction Coefficient: (E1%) E1%280 = 4.26
  • Solubility: Soluble in aqueous buffers (such as 10 mM Tris, pH 7.8 at 1 mg/mL).
  • CAS Number: 66676-43-5
  • Molecular Weight: 27,700 (sedimentation equilibrium)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Used for selective cleavage of Proteins for Amino Acid sequence determination or Peptide mapping
Staphylococcus aureus V8 Strain
Powder
Protease
4 and 7.8
-20°C
1 mg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Protease

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Protease

SDS
Documents

Documents

Certificates
Special Offers

Special Offers