Learn More
MP Biomedicals™ Protein Precipitate Solution (PPS)
Component of FastDNA™ SPIN Kit for Soil
Brand: MP Biomedicals™ 116540403
35.47 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
PPS is a lysis solution component included in the FastDNA and the FastDNA Spin Kits. Used as a stabilization agent during sample lysis of primarily plant tissue, but can also be used for other sample types that are high in inhibitory compounds. PPS is a protein precipitation solution used in conjunction with a the CLS-VF lysing buffer for homogenization of plant tissue for the subsequent extraction and purification of nucliec acids.
Specifications
Clear | |
FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-1 | |
6.18 | |
25 mL |
Stable at room temperature. Shelf life may be extended by storing at 4°C | |
Nucleic Acid Isolation & Purification | |
PPS | |
Liquid |