Learn More
MP Biomedicals™ RNAPro Solution
RNAPro Solution is a proprietary lysis buffer included with all of the FastRNA Pro purification kits. It can be used with the kits or with any other purification protocol as the primary lysis buffer to isolate total RNA from any type of plant or animal tissue, cultured cells, microbes, or soil and environmental samples.
Brand: MP Biomedicals™ 116055050
143.58 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 2922 Chem class : 8 Pack group : III
Description
- Extracted RNA is suitable for RT-PCR analysis and other downstream applications
- Using the FastPrep-24™ or FastPrep™ FP120 Instrument, samples can be homogenized by using MP Lysing Matrix tubes
- Use with FastPrep-24 or FastPrep-96 (with ConeFlex Legacy Adapter), or with optional CoolPrep Adapter kits to lyse, grind or homogenize samples, for the extraction and purification of RNA
Specifications
Room Temperature | |
FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1 | |
RNAPro Solution | |
Scalable |
Seeds, Wastewater, Bacteria (Gram +/-), Spores, Feces, Animal Tissues, Feces, Plants, Fungi, Soil, Roots, Soil | |
Isolation of RNA from most biological or environmental samples | |
2 x 27.5 mL |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : phenol, guanidine thiocyanate
Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.