Learn More
MP Biomedicals™ SC-HIS, Powder
Single dropout formulation of Synthetic Complete Supplement Mixture (SC) of amino acids for S. cereviseae growth media
Brand: MP Biomedicals™ 114410222
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Formulation
- Add 1.9g of SC-His to each liter of DOB or DOBA and autoclave at 121°C for 15 minutes for complete synthetic defined medium for S.cerevisiae
Yeast cell culture, Gene expression, Transcription regulation
Specifications
15°C to 30°C | |
Yeast Media | |
Yeast cell culture, gene expression, transcription regulation |
Powder | |
DOB or DOBA of 1.9g/L | |
30 g |