Learn More
MP Biomedicals™ SC-TRP, Powder
Single dropout formulation of Synthetic Complete Supplement Mixture (SC) of amino acids foS. cereviseae; growth media
Brand: MP Biomedicals™ 114410522
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Formulation
- Add 1.9g of SC-Trp to each liter of DOB or DOBA and autoclave at 121°C for 15 minutes for complete synthetic defined medium for S.cerevisiae
Applications:
Yeast cell culture, Gene expression, Transcription Regulation
Specifications
Molecular biology Certified™ | |
Powder | |
Adenine 21mg/L, L-Alanine 86.5mg/L, L-Arginine HCl 85.6mg/L, L-Asparagine 85.6mg/L, L-Aspartic Acid 85.6mg/L, L-Cysteine HCl 85.6mg/L, Glutamine 85.6mg/L, L-Glutamic Acid 85.6mg/L, Glycine 85.6mg/L, L-Histidine HCl 85.6mg/L, Myo-Inositol 85.6mg/L, L-Isoleucine 85.6mg/L, L-Leucine 173.4mg/L, L-Lysine HCl 85.6mg/L, L-Methionine 85.6mg/L, Para-Aminobenzoic Acid 8.6mg/L, L-Phenylalanine 85.6mg/L, L-Proline 85.6mg/L, L-Serine 85.6mg/L, L-Threonine 85.6mg/L, L-Tyrosine 85.6mg/L, Uracil 85.6mg/L, L-Valine 85.6mg/L | |
30 g |
15°C to 30°C | |
Yeast Media | |
For growth of Saccharomyces cerevisiae |