Learn More
MP Biomedicals™ Stem Cell Factor, Human, Recombinant
Stimulates the proliferation of bone marrow cells in 5-fluorouracil treated mice
Brand: MP Biomedicals™ 0215869802
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Hematopoietic growth factor, also known as Mast Cell Growth Factor or c-kit ligand
- Expressed in E. coli
- ED50 is 20.0 ng/mL
Believed to stimulate the proliferation of myeloid, erythroid and lymphoid progenitors in bone marrow cultures and acts synergistically with colony stimulating factors, resulting in greater size and yields
Order Info
Supplied lyophilized
Specifications
-20°C | |
Sterile filtered then the protein was lyophilized with no additives | |
≥98% | |
E. coli |
Stem Cell Factor | |
Believed to stimulate the proliferation of myeloid, erythroid and lymphoid progenitors in bone marrow cultures and acts synergistically with colony stimulating factors, resulting in greater size and yields | |
2μg | |
Human |