missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Streptokinase

Catalyzes hydrolysis of amide linkages

Brand:  MP Biomedicals™ 0210111410


 View more versions of this product

Product Code. 11328709

  • £49.80 / 10ku

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Forms a 1:1 complex with plasminogen which is then converted to streptokinase-plasmin. Both streptokinase-plasminogen and streptokinase-plasmin complexes can activate plasminogen.

  • Can be used as a thrombolytic agent in therapy of ischemic stroke
  • From: Lancefield group C strain of Streptococcus
  • Presentation: White lyophilized powder
  • Unit definition: Amount of enzyme that will liquefy a standard clot of fibrinogen, plasminogen and thrombin in 10 minutes at 37°C and pH 7.5
  • Specific activity: >5,000 units/mg dry weight
  • Solubility: Soluble in water

TRUSTED_SUSTAINABILITY
Specifications

Specifications

≥5000 U/mg
Used as a thrombolytic agent in the therapy of ischemic stroke
Streptococcus species, lancefield group C strain
Streptokinase
Lyophilized Powder
White
-20°C
10 kU
47400
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Streptokinase

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Streptokinase

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.