Learn More
MP Biomedicals™ TCM Defined Serum Replacement
Fortified, multipurpose serum replacement for the long-term culturing of anchorage dependent and suspension cells from a variety of species.
Brand: MP Biomedicals™ 092010026
198.38 GBP valid until 2024-06-28
Use promo code "21691" to get your promotional price.
Additional Details : Weight : 0.02000kg
Description
TCM is a fortified, multipurpose serum replacements for the long-term culturing of anchorage dependent and suspension cells from a variety of species. These serum replacements are effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.
- Universal serum replacement that supports human and mammalian cells
- Completely free of virus, mycoplasma and other infectious agents
- Contains no growth factors, steroid hormones or phenol red
- Highly purified and heat treated bovine albumin and transferrin are added with final total protein concentration less than 0.1%
- Packaged as a 50X concentrate; therefore, 10mL will produce 500mL of growth medium
Specifications
4°C | |
FBS Replacement Serum | |
20 mL | |
6.8 to 7.2 |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.