missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Terrific Broth II, Dehydrated Media
Rich medium for growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium
£104.00 - £954.00
Specifications
Certifications/Compliance | Molecular Biology Certified™ |
---|---|
Content And Storage | 15°C to 30°C |
Form | Powder |
Grade | Molecular Biology |
Product Type | Microbiology Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11367699
|
MP Biomedicals™
113046032 |
1000 g |
£235.00
1kg |
Estimated Shipment: 01-05-2025
Log in to see stock. |
||||||
11347699
|
MP Biomedicals™
113046022 |
454 g |
£104.00
454g |
Estimated Shipment: 01-05-2025
Log in to see stock. |
||||||
11387699
|
MP Biomedicals™
113046042 |
5000 g |
£954.00
5kg |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
Description
Specifications
Molecular Biology Certified™ | |
Powder | |
Microbiology Media | |
For growth of Escherichia coli |
15°C to 30°C | |
Molecular Biology | |
Yeast extract, Tryptone broth, Sodium Chloride and Vitamins |
Safety and Handling
missing translation for 'shelfLife' : 4 Years
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Terrific Broth II, Dehydrated Media