missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Tryptose Phosphate Broth
Provides suitable conditions for both aerobic growth in the clear upper zone and for microaerophilic and anaerobic growth in the lower, flocculent agar zones.
Brand: MP Biomedicals™ 091682149
This item is not returnable.
View return policy
Description
Specifications
15°C to 30°C | |
Tryptose Phosphate Broth | |
Tryptose 20g/L, Sodium Chloride 5g/L, Dextrose 2g/L, Disodium Phosphate 2.5g/L |
Liquid | |
Microbiology Media | |
100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Tryptose Phosphate Broth > Tryptose Phosphate Broth; 100mL
Spot an opportunity for improvement?Share a Content Correction