missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Wheat Germ Agglutinin
MP Biomedicals™ Wheat Germ Agglutinin has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers.
Brand: MP Biomedicals™ 0215226625
Code : NEW
Additional Details : Weight : 0.00010kg
Description
- Less than 4 mg/mL will agglutinate human type O erythrocytes, and less than 1 mg/mL will agglutinate neuraminidase-treated erythrocytes
- Contains no protein-bound carbohydrate
Specifications
White to Pale Yellow | |
Triticum vulgaris | |
Lyophilized | |
Lectin |
25 mg | |
-20°C | |
Proteomics, immune system regulation |