Learn More
MP Biomedicals™ Yeast Lytic Enzyme
From Arthrobacter luteus
Brand: MP Biomedicals™ 0215352610
Code : NEW
Additional Details : Weight : 1.36080kg
Description
Unit Definition: One unit will produce a ΔA800 of 0.001 per minute at pH 7.5 and 25°C, using a suspension of brewers yeast as a substrate in a 3 mL reaction mixture.
This enzyme catalyzes the lysis of yeast cells and hydrolyzes glucose polymers with β-1,3-linkages (e.g., glucan). It is also useful for preparing yeast spheroplasts.
Zymolyase
Lyophilized powder
Activity: 20,000 units/g
Specifications
20,000 U/g | |
7.5 | |
0°C | |
10 kU | |
Lyophilized Powder |
Tan | |
Digest the cell wall of active Yeast cells | |
Arthrobacter luteus | |
Yeast Lytic Enzyme |
Safety and Handling
- Yeast Lytic Enzyme, from arthrobacter luteus
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : yeast lytic enzyme