Learn More
MP Biomedicals™ YNB, Powdered Media: Without Ammonium Sulfate/Copper Sulfate, Ferric Chloride
Can be prepared by adding a carbon source, the appropriate amino acids, and agar if necessary
Brand: MP Biomedicals™ 114027112
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
- Nitrogen-based yeast
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Molecular Biology Certified™ | |
Powder | |
Vitamins: Biotin 0.002mg/L, Calcium Pantothenate 0.4mg/L, Folic Acid 0.002mg/L, Inositol 2mg/L, Niacin 0.4mg/L, P-Aminobenzoic Acid 0.2mg/L, Pyridoxine Hydrochloride 0.4mg/L, Riboflavin 0.2mg/L, Thiamine Hydrochloride 0.4mg/L; Trace Elements: Boric Acid 0.5mg, Potassium Iodide 0.1mg, Maganese Sulfate 0.4mg, Sodium Molybdate 0.2mg, Zinc Sulfate 0.4mg; Salts: Magnesium Sulfate 500mg, Sodium Chloride 100mg, Potassium Phosphate monobasic 1000mg, Calcium Chloride 100mg | |
227 g |
15°C to 30°C | |
Microbiology Media | |
For vigorous growth of Saccharomyces cerevisiae |
Safety and Handling
- YNB w/o Ammonium Sulfate, Copper Sulfate, Ferric Chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H360-May damage fertility or the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : p-Aminobenzoic Acid,Zinc Sulfate, Manganese Sulfate Monohydrate, Calcium Chloride Dihydrate, Potass