missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MP Magnetic Rack 8

This tool is for fast and easy extraction of nucleic acids using a magnetic method. It is commonly used in combination with MagBeads FastDNA™ kits.

Brand:  MP Biomedicals™ 116570426

Product Code. 17295114

  • £223.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Magnet made of high-energy neodymium
    • Small, strong and easy to handle
  • For fast and efficient magnetic separations
  • Design allows easy removal of rack from magnet
    • Tubes can be placed on shaker or vortex during extraction process
    • Good visibility of samples
  • Up to 8 tubes of 1 to 5 mL

Storage:

  • Magnet:
    • Stored in a dry and cool place
    • Never use > 50 °C
  • Tube rack:
    • Store at room temp
    • Can be placed with tubes at temp as low as -20°C for a limited time

Cleaning:

  • Use with one of the following solutions:
    • 70% (v/v) isopropyl alcohol
    • 1% (v/v) sodium hypochlorite solution (bleach)
    • 0.1 N HCl solution

Notes:

  • Avoid prolonged exposure of rack to water or aqueous solutions
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

8 x 1 to 5 mL tubes
Magnetic Rack
Magnetic Rack
Store magnet in dry, cool place, Store rack at room temp
For use with the Magbeads FastDNA™ kit, tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.