Learn More
MP Biomedicals™ MP-MRC-5™ Medium, Serum-free
Very low protein (90ng/mL) medium developed specifically for the culture of MRC-5 cells.
Brand: MP Biomedicals™ 092013426
Code : NEW
Additional Details : Weight : 1.00000kg
Description
MP-MRC-5 medium was developed specifically for microcarrier culture of MRC-5 cells in production systems. It is completely animal component-free and contains very little protein. MP-MRC-5 achieves growth rates superior to standard serum-supplemented media. Adaptation from serum-supplemented media is not necessary.
Recommended Storage
Store at +4°C. Protect from light. Do not freeze.
Specifications
Store at +4°C. Protect from light. Do not freeze. | |
MRC-5 cells | |
Liquid | |
Serum-free | |
Inorganic Salts, Essential and nonessential amino acids, Peptide supplements of plant origin, Vitamins, Growth Factors, Other organic compounds, Trace elements | |
Intended for use as a production medium to support the growth of MRC-5 cells in agitated (microcarrier) cultures | |
1 L |
MP-MRC-5 Medium | |
Very Low Protein (<90ng/mL) | |
Serum Free Medium (SFM) | |
Sterile | |
No Glutamine, No Animal Derived Components | |
7 |