missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPI 293T Cell Overexpression Lysate (Denatured), Abnova
Brand: Abnova H00004351-T01.100uL
Additional Details : Weight : 0.00010kg
Description
Western Blotting
Specifications
Western Blot | |
MPI 293T Cell Transient Overexpression Lysate(Denatured) | |
46.7 | |
pCMV-MPI full-length | |
100 μL | |
Human |
293T | |
NM_002435.1 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-MPI antibody (H00004351-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |