missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | MPI |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892774
|
Proteintech
14234-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882774
|
Proteintech
14234-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.Specifications
MPI | |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
P34949, Q68FX1, Q924M7 | |
110119, 300741, 4351 | |
MPI Fusion Protein Ag5475 | |
Primary | |
-20°C | |
MPI |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Mannose 6 phosphate isomerase, mannose phosphate isomerase, MPI, Phosphohexomutase, Phosphomannose isomerase, PMI, PMI1 | |
MPI | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |