missing translation for 'onlineSavingsMsg'
Learn More

MPP dihydrochloride, Tocris Bioscience™

Product Code. 18024832
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly selective ERα antagonist

Selective, high affinity silent antagonist at ERα receptors. Displays > 200-fold selectivity for ERα over ERβ. Ki values are 2.7 and 1800 nM at ERα and ERβ receptors respectively.

Chemical Identifiers

CAS 911295-24-4
Molecular Formula C29H33Cl2N3O3
Molecular Weight (g/mol) 542.501
InChI Key SMPRQALBFACGGO-UHFFFAOYSA-N
Synonym mpp dihydrochloride, 1,3-bis-4-hydroxyphenyl-4-methyl-5-4-2-piperidinylethoxy phenol-1h-pyrazole dihydrochloride, mpp dihydrochloride hydrate, methyl-piperidino-pyrazole hydrate, 1,3-bis 4-hydroxyphenyl-4-methyl-5-4-2-piperidinylethoxy phenol-1h-pyrazole dihydrochloride, mpp dihydrochloride hydrate hplc , white powder, 1,3-bis 4-hydroxyphenyl-4-methyl-5-4-2-piperidinylethoxy phenol-1h-pyrazole dihydrochloride hydrate, 4-1-4-hydroxyphenyl-4-methyl-5-4-2-piperidin-1-yl ethoxy phenyl-1,2-dihydro-3h-pyrazol-3-ylidene cyclohexa-2,5-dien-1-one-hydrogen chloride 1/2, 4-3-4-hydroxyphenyl-4-methyl-5-4-2-piperidin-1-yl ethoxy phenyl pyrazol-1-yl phenol dihydrochloride
PubChem CID 45073474
IUPAC Name 4-[2-(4-hydroxyphenyl)-4-methyl-3-[4-(2-piperidin-1-ylethoxy)phenyl]-1H-pyrazol-5-ylidene]cyclohexa-2,5-dien-1-one;dihydrochloride
SMILES CC1=C(N(NC1=C2C=CC(=O)C=C2)C3=CC=C(C=C3)O)C4=CC=C(C=C4)OCCN5CCCCC5.Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 542.5
Percent Purity >98%
Chemical Name or Material MPP dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'