missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MPure-12™ Automated Nucleic Acid Purification System

Purifies nucleic acids from a wide variety of bio-specimens using magnetic bead separation technology

Brand:  MP Biomedicals™ 117002200

Product Code. 16312116

  • £22875.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: MPure-12 Instrument with Bar Code Reader
This item is not returnable. View return policy

Description

Description

  • MPure-12 is a bench-top automated system
  • Purifies nucleic acids from a wide variety of bio-specimens using magnetic bead separation technology.
  • It has a uniquely designed magnetic bead processing chamber.
  • Fully integrated with easy to use pre-packaged reagent kits.

Certifications

CE

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Room Temperature
27.5 in.
70cm
Elution Volume: 50 to 300μL
1 to 12
Instruments and Systems
5 IU spiked (about 1 IU in PCR reaction)
110 to 240V
Made of Metal, Glass, Plastic
Magnetic Bead
Bacteria, Blood, Cell Cultures, FFPE, Forensic, Plants, Plasma, Serum, Swab, Tissue
Nucleic Acid Purification (DNA, RNA)
15°C to 30°C
100 to 1000μL of sample
Run Time: 30 to 70 minutes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers