Learn More
MP Biomedicals™ MPure™ Total RNA Extraction Kit
MPure™ Total RNA Extraction Kit used with the MPure-12 instrument for extraction of total RNA from whole blood, blood cells, animal tissue, plant tissue, yeast or cultured cells.
Brand: MP Biomedicals™ 117022160
307.80 GBP valid until 2025-12-31
Use promo code "24340" to get your promotional price.
This item is not returnable.
View return policy
Description
- Nucleic acids extracted can be used in a number of downstream applications including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP, Southern Blot Analysis.
Specifications
MPure-12 Total RNA Extraction Kit should be stored at room temperature (15°C to 25°C). Do not freeze the reagent cartridges. The kits are stable for 15 months under these conditions. | |
Whole Blood, Yeast, Cultured Cells, Animal Tissue, Plant Tissue, Blood Cells | |
Proteinase K Solution (40 μL), Lysis Buffer 2 (720 μL), Binding Buffer 1 (1300 μL), Magnetic Bead Solution (800 μL), Washing Buffer 1 (1000 μL), Washing Buffer 2 (1000 μL), Washing Buffer 3 (1000 μL), RNase Free Water (1000 μL), Rnase Free Water (1000 μL), DNase | |
48 Tests | |
MPure Total RNA Extraction Kit |
Kit | |
RT-PCR, qPCR, sequencing (NGS), microarray, DDRT PCR, TaqMan™ analysis, northern blot analysis, northern blotting | |
RNA Extraction Kit | |
Animal and Human Tissues |
Product Content Correction
Product Title
MP Biomedicals™ MPure™ Total RNA Extraction Kit > MPure™ Total RNA Extraction Kit; No. of Reactions: 48 Tests
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Comparta una corrección de contenido